missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Cardiotrophin-1/CT-1 Recombinant Protein

$378.79 - $1,176.89

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 1489
Molecular Weight 21 kDa
Name Cardiotrophin-1/CT-1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
612CD010 R&D Systems
RDS612CD010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $378.79
612CD050 R&D Systems
RDS612CD050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $1,176.89
612CD010CF R&D Systems
RDS612CD010CF
Supplied as a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $378.79
612CD050CF R&D Systems
RDS612CD050CF
Supplied as a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $1,176.89
Specifications

Description

Source: E. coli-derived, Ser2-Ala201
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Supplied as a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 1489
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
21 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
1489
Cardiotrophin-1/CT-1
Yes