missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human C-Reactive Protein/CRP Recombinant Protein

$311.55 - $311.55

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 1401
Molecular Weight 23 kDa (monomer)
Name C-Reactive Protein/CRP
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1707CR200 R&D Systems
RDS1707CR200
Lyophilized from a 0.2μm filtered solution in Tris-HCl, NaCl and CaCl2 with BSA as a carrier protein. Each for $311.55
1707CR200CF R&D Systems
RDS1707CR200CF
Lyophilized from a 0.2μm filtered solution in Tris-HCl, NaCl and CaCl2. Each for $311.55
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Phe17-Pro224
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 23 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-HCl, NaCl and CaCl2.
Entrez Gene ID: 1401
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
23 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
1401
C-Reactive Protein/CRP
200μg
Human