Please login to your online account to display your discounted pricing

R&D Systems Human BMP-9 Recombinant Protein

$521.89 - $521.89

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2658
Molecular Weight 12.1 kDa (monomer)
Name BMP-9
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

3209BP010CF

 
r&d systems
RDS3209BP010CF
Supplied as a 0.2μm filtered solution in Acetonitrile and TFA. Each for $521.89

3209BP010

 
r&d systems
RDS3209BP010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $521.89
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2658
Molecular Weight 12.1 kDa (monomer)
Name BMP-9
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Human

Source: Chinese Hamster Ovary cell line, CHO-derived, Ser320-Arg429
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.1 kDa (monomer)
Formulation: Supplied as a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 2658
Regulatory Status: RUO