missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human BMP-8a Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 656
Molecular Weight 15.8 kDa (monomer)
Name BMP-8a
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1073BP010 R&D Systems
RDS1073BP010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $376.73
1073BP010CF R&D Systems
RDS1073BP010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $376.73
Specifications

Description

Source: E. coli-derived, Ala264-His402 & Val265-His402
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.8 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 656
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.8 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
656
BMP-8a
10μg
Human