Please login to your online account to display your discounted pricing

R&D Systems Human BMP-4 Recombinant Protein

$475.80 - $1,250.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 652
Molecular Weight 13 kDa (monomer)
Name BMP-4
Purity 95%, by SDS-PAGE with silver staining.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

314BP010

 
r&d systems
RDS314BP010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $475.80

314BP050

 
r&d systems
RDS314BP050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $1,250.12

314BP010CF

 
r&d systems
RDS314BP010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $475.80

314BP050CF

 
r&d systems
RDS314BP050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $1,250.12
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 652
Molecular Weight 13 kDa (monomer)
Name BMP-4
Purity 95%, by SDS-PAGE with silver staining.
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Ser293-Arg408
Purity: 95%, by SDS-PAGE with silver staining.
Molecular Weight: 13 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 652
Regulatory Status: RUO