Please login to your online account to display your discounted pricing

R&D Systems Human BMP-4/BMP-7 Heterodimer Recombinant Protein

$521.43 - $521.43

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 652
Molecular Weight 13.3 kDa (BMP-4), 15.8 kDa (BMP-7)
Name BMP-4/BMP-7 Heterodimer
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

3727BP010CF

 
r&d systems
RDS3727BP010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $521.43

3727BP010

 
r&d systems
RDS3727BP010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $521.43
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 652
Molecular Weight 13.3 kDa (BMP-4), 15.8 kDa (BMP-7)
Name BMP-4/BMP-7 Heterodimer
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Human

Source: E. coli-derived, Ser293-Arg408
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.3 kDa (BMP-4), 15.8 kDa (BMP-7)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 652
Regulatory Status: RUO