missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human BMP-3b/GDF-10 Recombinant Protein

$518.14 - $518.14

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2662
Molecular Weight 12.5 kDa (monomer)
Name BMP-3b/GDF-10
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1543BP025 R&D Systems
RDS1543BP025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $518.14
1543BP025CF R&D Systems
RDS1543BP025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $518.14
Specifications

Description

Source: E. coli-derived, Gln369-Arg478, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.5 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 2662
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
12.5 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
2662
BMP-3b/GDF-10
25μg
Human