missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human BMP-3 Recombinant Protein

$527.94 - $527.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 651
Molecular Weight 12.5 kDa (monomer)
Name BMP-3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
113BP100CF R&D Systems
RDS113BP100CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $527.94
113BP100 R&D Systems
RDS113BP100
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $527.94
Specifications

Description

Source: E. coli-derived, Gln363-Arg472, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.5 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 651
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
12.5 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
651
BMP-3
100μg
Human