missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human BMP-2 Recombinant Protein, (E. coli-expressed)

For use in cell or tissue culture applications.

$272.44 - $272.44

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 650
Molecular Weight 12.8 kDa (monomer)
Name BMP-2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
355BEC010 R&D Systems
RDS355BEC010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $272.44
355BEC010CF R&D Systems
RDS355BEC010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $272.44
Specifications

Description

  • Source: E. coli-derived, Ala284-Arg396
  • Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular Weight: 12.8 kDa (monomer)
  • Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
  • Entrez Gene ID: 650
  • Regulatory Status: RUO
Description & Specifications

Specifications

Unlabeled
12.8 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
Disulfide-linked homodimer
650
BMP-2
10μg
Human