missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human BMP-2/BMP-6 Heterodimer Recombinant Protein

$506.74 - $506.74

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 650
Molecular Weight 12.8 kDa (BMP-2) & 15 kDa (BMP-6)
Name BMP-2/BMP-6 Heterodimer
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7145BP010CF R&D Systems
RDS7145BP010CF
Lyophilized from a 0.2μm filtered solution in HCl. Each for $506.74
7145BP010 R&D Systems
RDS7145BP010
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. Each for $506.74
Specifications

Description

Source: E. coli-derived, Ala284-Arg396 (BMP-2) with an N-terminal Met & Gln382-His513 (BMP-6), with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.8 kDa (BMP-2) & 15 kDa (BMP-6)
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Entrez Gene ID: 650
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
12.8 kDa (BMP-2) & 15 kDa (BMP-6)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
650
BMP-2/BMP-6 Heterodimer
10μg
Human