missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human BMP-10 Recombinant Protein

$637.44 - $637.44

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 27302
Molecular Weight 12.2 kDa (monomer)
Name BMP-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2926BP025 R&D Systems
RDS2926BP025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $637.44
2926BP025CF R&D Systems
RDS2926BP025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $637.44
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Asn317-Arg424
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.2 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 27302
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
12.2 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
27302
BMP-10
25μg
Human