missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human BAFF/BLyS/TNFSF13B Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 10673
Molecular Weight 19.6 kDa
Name BAFF/BLyS/TNFSF13B
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2149BF010 R&D Systems
RDS2149BF010
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein. Each for $298.51
2149BF010CF R&D Systems
RDS2149BF010CF
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT. Each for $298.51
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ala134-Leu285
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT.
Entrez Gene ID: 10673
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
19.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
10673
BAFF/BLyS/TNFSF13B
10μg
Human