missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human B7-2/CD86 Fc Chimera Recombinant Protein

$428.87 - $441.90

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 942
Name B7-2/CD86 Fc Chimera
Quantity 100μg
Recombinant Yes
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Purity Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Purity Price Quantity  
141B2100 R&D Systems
RDS141B2100
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. 52.7 kDa (monomer) 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. Each for $441.90
7625B2100 R&D Systems
RDS7625B2100
Lyophilized from a 0.2μm filtered solution in PBS. 52 kDa (monomer) 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. Each for $428.87
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Leu20-His239
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 52 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 942
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
B7-2/CD86 Fc Chimera
Yes
942
100μg
Human