missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human APRIL/TNFSF13 Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8741
Molecular Weight 21.7 kDa
Name APRIL/TNFSF13
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5860AP010 R&D Systems
RDS5860AP010
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protin. Each for $415.83
5860AP010CF R&D Systems
RDS5860AP010CF
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl. Each for $415.83
Specifications

Description

Source: Human embryonic kidney cell, HEK293-derived, Ala105-Leu250
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Entrez Gene ID: 8741
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
21.7 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
8741
APRIL/TNFSF13
10μg
Human