Please login to your online account to display your discounted pricing

R&D Systems Human Angiopoietin-2 Recombinant Protein


Manufacturer: r&d systems  RDSBT623

 View more versions of this product

Catalog No. BT623

  • / Each
 

Description & Specifications

Specifications

Name Angiopoietin-2
Quantity 10μg
Recombinant Yes
Source Human
Structural Form Oligomer
Conjugate Biotin
Formulation Lyophilized from a 0.2μm filtered solution in Tris, NaCl and CHAPS with BSA as a carrier protein.
Gene ID (Entrez) 285