missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Angiopoietin-1 Recombinant Protein

$394.98 - $394.98

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 284
Molecular Weight 56 kDa
Name Angiopoietin-1
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
923AN025 R&D Systems
RDS923AN025
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl with BSA as a carrier protein. Each for $394.98
923AN025CF R&D Systems
RDS923AN025CF
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl. Each for $394.98
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ser20-Phe498, with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 56 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl.
Entrez Gene ID: 284
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
56 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
284
Angiopoietin-1
25μg
Human