missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Amphiregulin Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 374
Molecular Weight 11 kDa
Name Amphiregulin
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
262AR100 R&D Systems
RDS262AR100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
262AR100CF R&D Systems
RDS262AR100CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: E. coli-derived, Ser101-Lys198
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 11 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 374
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
11 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
374
Amphiregulin
100μg
Human