missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Activin RIIB Fc Chimera Recombinant Protein, (NS0-expressed)

$429.46 - $429.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 93
Molecular Weight 41 kDa (monomer)
Name Activin RIIB Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
339RB100 R&D Systems
RDS339RB100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $429.46
339RB100CF R&D Systems
RDS339RB100CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $429.46
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ser19-Thr134
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 41 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 93
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
41 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
93
Activin RIIB Fc Chimera
100μg
Human