missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Activin RIIA Fc Chimera Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 92
Molecular Weight 40 kDa (monomer)
Name Activin RIIA Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
340R2100CF R&D Systems
RDS340R2100CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
340R2100 R&D Systems
RDS340R2100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ser25-Pro134
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 40 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 92
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
40 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
92
Activin RIIA Fc Chimera
100μg
Human