Please login to your online account to display your discounted pricing

R&D Systems Human Activin B Recombinant Protein

$330.49 - $976.15

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3625
Molecular Weight 12.8 kDa (monomer)
Name Activin B
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

659AB005

 
r&d systems
RDS659AB005
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 5μg Each for $330.49

659AB005CF

 
r&d systems
RDS659AB005CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose. 5μg Each for $330.49

659AB025

 
r&d systems
RDS659AB025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 25μg Each for $976.15

659AB025CF

 
r&d systems
RDS659AB025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose. 25μg Each for $976.15
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3625
Molecular Weight 12.8 kDa (monomer)
Name Activin B
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: Chinese Hamster Ovary cell line, CHO-derived, Gly293-Ala407
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.8 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose.
Entrez Gene ID: 3625
Regulatory Status: RUO