missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Activin AC Recombinant Protein

$441.90 - $441.90

Specifications

Conjugate Unlabeled
Molecular Weight 12.9 kDa (betaA subunit), 12.5 kDa (betaC subunit)
Name Activin AC
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
4879AC010 R&D Systems
RDS4879AC010
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. Each for $441.90
4879AC010CF R&D Systems
RDS4879AC010CF
Lyophilized from a 0.2μm filtered solution in HCl. Each for $441.90
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Gly311-Ser426 (beta A) Gly237-Ser352 (beta C)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.9 kDa (betaA subunit), 12.5 kDa (betaC subunit)
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
Activin AC
10μg
Human
12.9 kDa (betaA subunit), 12.5 kDa (betaC subunit)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes