Please login to your online account to display your discounted pricing

R&D Systems Human Activin AB Recombinant Protein


Manufacturer: r&d systems  RDS1066AB005CF

 View more versions of this product

Catalog No. 1066AB005CF

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 12.8 kDa (betaA subunit) and 13 kDa (betaB subunit)
Name Activin AB
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 5μg
Recombinant Yes
Source Human
Structural Form Disulfide-linked heterodimer
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose.