missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Active Caspase-2 Recombinant Protein

$467.46 - $467.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 835
Molecular Weight 12 kDa & 18 kDa
Name Active Caspase-2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
702C2010CF R&D Systems
RDS702C2010CF
Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose. Each for $467.46
Specifications

Description

Source: E. coli-derived, Gly170-Asp333 & Ala348-Tyr452
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12 kDa & 18 kDa
Formulation: Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose.
Entrez Gene ID: 835
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
12 kDa & 18 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
835
Active Caspase-2
10μg
Human