missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human 4-1BB Ligand/TNFSF9 Recombinant Protein

$350.65 - $350.65

Specifications

Gene ID (Entrez) 8744
Host Species Human
Molecular Weight 20.4 kDa
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
22954L025CF R&D Systems
RDS22954L025CF
Lyophilized from a 0.2μm filtered solution in PBS, NaCl and PEG. Each for $350.65
22954L025 R&D Systems
RDS22954L025
Lyophilized from a 0.2μm filtered solution in PBS, NaCl and PEG with BSA as a carrier protein. Each for $350.65
Specifications

Description

Source: E. coli-derived, Arg71-Glu254, with an N-terminal Met and a 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, NaCl and PEG.
Entrez Gene ID: 8744
Regulatory Status: RUO

Description & Specifications

Specifications

8744
20.4 kDa
25μg
Human
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes