missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human 26S Proteasome Protein

$290.69 - $462.50

Specifications

Formulation Supplied as a solution in HEPES, NaCl, MgATP and Glycerol.
Host Species Human
Molecular Weight 2,500 kDa
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E365025 R&D Systems
RDSE365025
25μg Each for $290.69
E365050 R&D Systems
RDSE365050
50μg Each for $462.50
Specifications

Description

Source: Human embryonic kidney cell, HEK293-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 2,500 kDa
Formulation: Supplied as a solution in HEPES, NaCl, MgATP and Glycerol.
Regulatory Status: RUO

Description & Specifications

Specifications

Supplied as a solution in HEPES, NaCl, MgATP and Glycerol.
Human
2,500 kDa
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.