Please login to your online account to display your discounted pricing

R&D Systems Human 26S Proteasome Protein


Manufacturer: r&d systems  RDSE365025

 View more versions of this product

Catalog No. E365025

  • / Each
 

Description & Specifications

Specifications

Product Type 26S Proteasome
Host Species Human
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight 2,500 kDa
Quantity 25μg
Formulation Supplied as a solution in HEPES, NaCl, MgATP and Glycerol.