missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Guinea Pig TNF-alpha Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Molecular Weight 17.4 kDa
Name TNF-alpha
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5035TG025CF R&D Systems
RDS5035TG025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
5035TG025 R&D Systems
RDS5035TG025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: E. coli-derived, Leu79-Leu234, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
TNF-alpha
25μg
Guinea Pig
17.4 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes