missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Guinea Pig IL-10 Recombinant Protein

$389.76 - $389.76

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 100135553
Molecular Weight 18.5 kDa (monomer)
Name IL-10
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5258GL010CF R&D Systems
RDS5258GL010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $389.76
5258GL010 R&D Systems
RDS5258GL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $389.76
Specifications

Description

Source: E. coli-derived, Ser19-Ser178
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.5 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 100135553
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18.5 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
100135553
IL-10
10μg
Guinea Pig