missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Feline TNF-alpha Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493755
Molecular Weight 17.3 kDa
Name TNF-alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2586FT025 R&D Systems
RDS2586FT025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
2586FT025CF R&D Systems
RDS2586FT025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: E. coli-derived, Leu77-Leu233 (Thr210Ala), with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 493755
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
493755
TNF-alpha
25μg
Feline