Please login to your online account to display your discounted pricing

R&D Systems Feline IL-6 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493687
Molecular Weight 20.7 kDa
Name IL-6
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

2305FL025

 
r&d systems
RDS2305FL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

2305FL025CF

 
r&d systems
RDS2305FL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493687
Molecular Weight 20.7 kDa
Name IL-6
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Feline

Source: E. coli-derived, Thr28-Met208 (Glu133Lys), with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 493687
Regulatory Status: RUO