missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Feline IL-5 Recombinant Protein

$311.55 - $311.55

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493803
Molecular Weight 14.6 kDa
Name IL-5
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1795FL010CF R&D Systems
RDS1795FL010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $311.55
1795FL010 R&D Systems
RDS1795FL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $311.55
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ile20-Ser134, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 493803
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
14.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
493803
IL-5
10μg
Feline