Please login to your online account to display your discounted pricing

R&D Systems Feline IL-4 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Molecular Weight 12.6 kDa
Name IL-4
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

984FL025

 
r&d systems
RDS984FL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

984FL025CF

 
r&d systems
RDS984FL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 12.6 kDa
Name IL-4
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Feline

Source: E. coli-derived, Gly24-His133 & Asn26-His133
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO