Please login to your online account to display your discounted pricing

R&D Systems Feline IL-2 (Cys146Ser) Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 751114
Molecular Weight 15.6 kDa
Name IL-2 (Cys146Ser)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1890FL020CF

 
r&d systems
RDS1890FL020CF
Lyophilized from a 0.2μm filtered solution in Sodium Acetate. Each for $428.87

1890FL020

 
r&d systems
RDS1890FL020
Lyophilized from a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 751114
Molecular Weight 15.6 kDa
Name IL-2 (Cys146Ser)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 20μg
Recombinant Yes
Source Feline

Source: E. coli-derived, Ala21-Thr154 (Cys146Ser), with and without an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Acetate.
Entrez Gene ID: 751114
Regulatory Status: RUO