missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Feline IL-18/IL-1F4 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Molecular Weight 18.3 kDa
Name IL-18/IL-1F4
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5989RM025CF R&D Systems
RDS5989RM025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT. Each for $416.79
5989RM025 R&D Systems
RDS5989RM025
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: E. coli-derived, Tyr36-Asn192, with an N-terminal Met
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-18/IL-1F4
25μg
Feline
18.3 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes