Please login to your online account to display your discounted pricing

R&D Systems Feline IL-15 Recombinant Protein

$368.77 - $368.77

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493682
Molecular Weight 13 kDa
Name IL-15
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

7805FL010

 
r&d systems
RDS7805FL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $368.77

7805FL010CF

 
r&d systems
RDS7805FL010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $368.77
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493682
Molecular Weight 13 kDa
Name IL-15
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Feline

Source: E. coli-derived, Asn49-Ser162
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 493682
Regulatory Status: RUO