Please login to your online account to display your discounted pricing

R&D Systems Feline IL-10 Recombinant Protein

$389.76 - $389.76

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493683
Molecular Weight 18.9 kDa
Name IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

736FL010CF

 
r&d systems
RDS736FL010CF
Lyophilized from a 0.2μm filtered solution in NaH2PO4. Each for $389.76

736FL010

 
r&d systems
RDS736FL010
Lyophilized from a 0.2μm filtered solution in NaH2PO4 with BSA as a carrier protein. Each for $389.76
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493683
Molecular Weight 18.9 kDa
Name IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Feline

Source: E. coli-derived, Ser19-Ile178 & Ser26-Ile178
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4.
Entrez Gene ID: 493683
Regulatory Status: RUO