Please login to your online account to display your discounted pricing

R&D Systems Feline Flt-3 Ligand Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493796
Molecular Weight 18.5 kDa
Name Flt-3 Ligand
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

2467FL025

 
r&d systems
RDS2467FL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81

2467FL025CF

 
r&d systems
RDS2467FL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493796
Molecular Weight 18.5 kDa
Name Flt-3 Ligand
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Feline

Source: Mouse myeloma cell line, NS0-derived, Ser27-Pro185, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 493796
Regulatory Status: RUO