Please login to your online account to display your discounted pricing

R&D Systems Feline CXCL8/IL-8 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493836
Molecular Weight 9 kDa
Name CXCL8/IL-8
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

2277FL025

 
r&d systems
RDS2277FL025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $428.87

2277FL025CF

 
r&d systems
RDS2277FL025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493836
Molecular Weight 9 kDa
Name CXCL8/IL-8
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Feline

Source: E. coli-derived, Ala23-Ala101
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 493836
Regulatory Status: RUO