missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Feline CCL5/RANTES Recombinant Protein

$363.69 - $363.69

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 493689
Molecular Weight 7.7 kDa
Name CCL5/RANTES
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3819FR025 R&D Systems
RDS3819FR025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $363.69
3819FR025CF R&D Systems
RDS3819FR025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $363.69
Specifications

Description

Source: E. coli-derived, Tyr26-Asn92 (Lys84Tyr and Gly85 del)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 493689
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
493689
CCL5/RANTES
25μg
Feline