missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Equine TNF-alpha Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 100033834
Molecular Weight 17.2 kDa
Name TNF-alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1814ET025 R&D Systems
RDS1814ET025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
1814ET025CF R&D Systems
RDS1814ET025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: E. coli-derived, Leu78-Leu234, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 100033834
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
100033834
TNF-alpha
25μg
Equine