Please login to your online account to display your discounted pricing

R&D Systems Equine IL-6 Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Molecular Weight 21 kDa
Name IL-6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1886EL025

 
r&d systems
RDS1886EL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81

1886EL025CF

 
r&d systems
RDS1886EL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 21 kDa
Name IL-6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Equine

Source: E. coli-derived, Phe26-Met208, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO