missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Equine IL-4 Recombinant Protein

$441.90 - $441.90

Specifications

Conjugate Unlabeled
Molecular Weight 12.6 kDa
Name IL-4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 20μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1809EL020 R&D Systems
RDS1809EL020
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $441.90
1809EL020CF R&D Systems
RDS1809EL020CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $441.90
Specifications

Description

Source: E. coli-derived, Lys26-Cys137
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-4
20μg
Equine
12.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes