missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Equine IL-2 (Cys141Ser) Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 100034204
Molecular Weight 15 kDa
Name IL-2 (Cys141Ser)
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1613IL020 R&D Systems
RDS1613IL020
Lyophilized from a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein. Each for $428.87
1613IL020CF R&D Systems
RDS1613IL020CF
Lyophilized from a 0.2μm filtered solution in Sodium Acetate. Each for $428.87
Specifications

Description

Source: E. coli-derived, Ala21-Thr149 (Cys141Ser), with and without an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Acetate.
Entrez Gene ID: 100034204
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
100034204
IL-2 (Cys141Ser)
20μg
Equine