missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Equine IL-10 Recombinant Protein

$389.76 - $389.76

Specifications

Conjugate Unlabeled
Molecular Weight 19 kDa
Name IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1605IL010CF R&D Systems
RDS1605IL010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $389.76
1605IL010 R&D Systems
RDS1605IL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $389.76
Specifications

Description

Source: E. coli-derived, Ser19-Asn178
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-10
10μg
Equine
19 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes