missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Equine IL-1 beta/IL-1F2 Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 100034237
Molecular Weight 17 kDa
Name IL-1 beta/IL-1F2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3340EL010CF R&D Systems
RDS3340EL010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $376.73
3340EL010 R&D Systems
RDS3340EL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $376.73
Specifications

Description

Source: E. coli-derived, Ala116-Ala268 (Glu179Gly, Met188Thr, Thr194Ile, Ser245Lys and Arg256Gln)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 100034237
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
100034237
IL-1 beta/IL-1F2
10μg
Equine