missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Equine IFN-gamma Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Molecular Weight 16.7 kDa
Name IFN-gamma
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1586HG025 R&D Systems
RDS1586HG025
Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween™ 80 with BSA as a carrier protein. Each for $376.73
1586HG025CF R&D Systems
RDS1586HG025CF
Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween™ 80. Each for $376.73
Specifications

Description

Source: E. coli-derived, Ala25-Gln166
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween™ 80.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IFN-gamma
25μg
Equine
16.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes