Please login to your online account to display your discounted pricing

R&D Systems Drosophila Decapentaplegic/DPP Recombinant Protein

$458.10 - $458.10

Specifications

Conjugate Unlabeled
Molecular Weight 14.8 kDa (monomer)
Name Decapentaplegic/DPP
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 20μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

159DP020CF

 
r&d systems
RDS159DP020CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $458.10

159DP020

 
r&d systems
RDS159DP020
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $458.10
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 14.8 kDa (monomer)
Name Decapentaplegic/DPP
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 20μg
Recombinant Yes
Source Drosophila

Source: E. coli-derived, Asp457-Arg588 (Gln473His) & (Pro474Ala), with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.8 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO