Please login to your online account to display your discounted pricing

R&D Systems d-Luciferin Recombinant Protein

$31.90 - $292.21

Specifications

Conjugate Unlabeled
Name d-Luciferin
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

800LN01M

 
r&d systems
RDS800LN01M
1mg Each for $31.90

800LN05M

 
r&d systems
RDS800LN05M
5mg Each for $70.18

800LN50M

 
r&d systems
RDS800LN50M
50mg Each for $292.21
Description & Specifications

Specifications

Conjugate Unlabeled
Name d-Luciferin
Recombinant Yes

Source: Chemically Synthesized
Regulatory Status: RUO