Please login to your online account to display your discounted pricing
R&D Systems CultreCoat™ BME Cell Invasion Assays, 96 well
Reagent kit for evaluating cell invasion for low, intermediary or highly invasive cell models
$521.43 - $521.43
Specifications
Quantity 1 Kit
View More Specs
Includes:  96 Well BME Cell Invasion Chamber (Low, Medium or High); 25X Cell Wash Buffer, 2 x 1.5mL; 10X Cell Dissociation Solution, 2 x 1.5mL; Calcein-AM, 50ug
Products
Catalog Number Mfr. No. Type Price Quantity    
3481096K
r&d systems
3481096K
Low BME Cell Invasion Assay Each for $521.43
3482096K
r&d systems
3482096K
Medium BME Cell Invasion Assay Each for $521.43
3483096K
r&d systems
3483096K
High BME Cell Invasion Assay Each for $521.43
Description & Specifications
Specifications
Quantity 1 Kit
Cell invasion is fundamental to angiogenesis, embryonic development, immune responses and tumor cell metastasis.
Contains one 96-well plate that is coated with BME, for cell models that exhibit a nominal invasive capacity: The low amount of BME coating provides the highest sensitivity for minimally invasive cell models
The medium amount of BME coating provides the