missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Cotton Rat TNF-alpha Recombinant Protein

$272.44 - $272.44

Specifications

Conjugate Unlabeled
Molecular Weight 17.5 kDa
Name TNF-alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1011CR025CF R&D Systems
RDS1011CR025CF
Lyophilized from a 0.2μm filtered solution in NaH2PO4, NaCl and EDTA. Each for $272.44
1011CR025 R&D Systems
RDS1011CR025
Lyophilized from a 0.2μm filtered solution in NaH2PO4, NaCl and EDTA with BSA as a carrier protein. Each for $272.44
Specifications

Description

Source: E. coli-derived, Leu1-Leu156, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4, NaCl and EDTA.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
TNF-alpha
25μg
Cotton Rat
17.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes